n-Butyl levulinate, 98%
n-Butyl levulinate, 98%
n-Butyl levulinate, 98%
n-Butyl levulinate, 98%
Thermo Scientific Chemicals
n-Butyl levulinate, 98%
CAS: 2052-15-5 | C9H16O3 | 172.224 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFH59274.AP
Price (MYR)
552.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
552.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Butyl levulinate
CAS2052-15-5
Health Hazard 1H227
Health Hazard 3P210-P280-P370+P378q-P501c
Recommended StorageAmbient temperatures
View more
Butyl levulinate was used in the synthesis of γ-valerolactone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Butyl levulinate was used in the synthesis of γ-valerolactone.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Xian-Long Du; Qing-Yuan Bi; Yong-Mei Liu; Yong Cao; Kang-Nian Fan. Conversion of biomass-derived levulinate and formate esters into γ-valerolactone over supported gold catalysts. Chemsuschem Chemistry And Sustainability, Energy & Materials,2011,4(12), 1838-1843.
  2. S. Dharne; V.V. Bokade. Esterification of levulinic acid to n-butyl levulinate over heteropolyacid supported on acid-treated clay. Journal of Natural Gas Chemistry,2011,20(1), 18-24.