4-Nitrothiophenol, 96%
4-Nitrothiophenol, 96%
4-Nitrothiophenol, 96%
4-Nitrothiophenol, 96%
Thermo Scientific Chemicals
4-Nitrothiophenol, 96%
CAS: 1849-36-1 | C6H5NO2S | 155.171 g/mol
Have Questions?
Catalog NumberQuantity
ALFH59240.065 g
Catalog number ALFH59240.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitrothiophenol
CAS1849-36-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Partly soluble in water and chloroform.
Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Jens U. Nielsen.; Maria J. Esplandiu.; Dieter M. Kolb. 4-Nitrothiophenol SAM on Au(111) Investigated by in Situ STM, Electrochemistry, and XPS. Langmuir. 2001, 17 (11), 3454-3459.
  2. Hiromori Tsutsumi.; Shozo Furumoto.; Masayuki Morita.; Yoshiharu Matsuda. Electrochemical Behavior of a 4-Nitrothiophenol Modified Electrode Prepared by the Self-Assembly Method. J. Colloid Interface Sc. 1995, 171 (2),505-511.