2-n-Propyl-1-pentanol, 97%
2-n-Propyl-1-pentanol, 97%
2-n-Propyl-1-pentanol, 97%
2-n-Propyl-1-pentanol, 97%
Thermo Scientific Chemicals
2-n-Propyl-1-pentanol, 97%
CAS: 58175-57-8 | C8H18O | 130.231 g/mol
Have Questions?
Catalog NumberQuantity
ALFH57954.031 g
Catalog number ALFH57954.03
Price (MYR)
330.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
330.00
EA
Add to cart
Specifications
Chemical Name or Material2-n-Propyl-1-pentanol
CAS58175-57-8
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c