(+)-Usnic acid, 98%
(+)-Usnic acid, 98%
(+)-Usnic acid, 98%
Thermo Scientific Chemicals
(+)-Usnic acid, 98%
CAS: 7562-61-0 | C18H16O7 | 344.32 g/mol
Have Questions?
Catalog NumberQuantity
ALFH56612.065 g
Catalog number ALFH56612.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material(+)-Usnic acid
CAS7562-61-0
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
It is used as primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as primary and secondary intermediate.
Solubility
Partly soluble in water. Soluble in acetone, ethanol, chloroform and furfural.
Notes
Store at 4°C. Reactive with oxidizing agents.
RUO – Research Use Only
General References:
  1. K. Ingólfsdóttir. Usnic acid. Phytochemistry. 2002, 61 (7),729-736.
  2. Moreno Cocchietto.; Nicola Skert.; Pier Nimis.; Gianni Sava. A review on usnic acid, an interesting natural compound. Naturwissenschaften. 2002, 89 (4),137-146.