1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
Thermo Scientific Chemicals
1,10-Phenanthroline-5,6-dione, 98%
CAS: 27318-90-7 | C12H6N2O2 | 210.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFH55747.031 g
Catalog number ALFH55747.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material1,10-Phenanthroline-5,6-dione
CAS27318-90-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.
Solubility
Insoluble in water.
Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
RUO – Research Use Only
General References:
  1. Charles A. Goss.; Hector D. Abruna. Spectral, electrochemical and electrocatalytic properties of 1,10-phenanthroline-5,6-dione complexes of transition metals. Inorg. Chem. 1985, 24 (25), 4263-4267.
  2. Q. Wu.; M. Maskus.; F. Pariente.; F. Tobalina.; V. M. Fernández.; E. Lorenzo.; H. D. Abruña. Electrocatalytic Oxidation of NADH at Glassy Carbon Electrodes Modified with Transition Metal Complexes Containing 1,10-Phenanthroline-5,6-dione Ligands. Anal. Chem. 1996, 68 (20),3688-3696.