Tris(8-hydroxyquinolinato)aluminum, 99%
Tris(8-hydroxyquinolinato)aluminum, 99%
Tris(8-hydroxyquinolinato)aluminum, 99%
Tris(8-hydroxyquinolinato)aluminum, 99%
Thermo Scientific Chemicals
Tris(8-hydroxyquinolinato)aluminum, 99%
CAS: 2085-33-8
Have Questions?
Catalog NumberQuantity
ALFH55656.065 g
Catalog number ALFH55656.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTris(8-hydroxyquinolinato)aluminum
CAS2085-33-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Key green-light emitting and electron transport material for OLEDs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Key green-light emitting and electron transport material for OLEDs.
Solubility
Solubility in chloroform
Notes
Reacts with strong oxidizing agents. Store at room temperature.
RUO – Research Use Only
General References:
  1. Bo Chao Lin.; Cheu P. Cheng.; Zhi-Qiang You.; Chao-Ping Hsu. Charge Transport Properties of Tris(8-hydroxyquinolinato)aluminum(III): Why It Is an Electron Transporter.J. Am. Chem. Soc. 2005, 127 (1), 66-67.
  2. Gary P. Kushto.; Yasuhiro Iizumi.; Junji Kido.; Zakya H. Kafafi. A Matrix-Isolation Spectroscopic and Theoretical Investigation of Tris(8-hydroxyquinolinato)aluminum(III) and Tris(4-methyl-8-hydroxyquinolinato)aluminum(III).J. Phys. Chem. 2000, 104 (16),3670-3680.