3,3'-Diethyloxacarbocyanine iodide, 98%
3,3'-Diethyloxacarbocyanine iodide, 98%
3,3'-Diethyloxacarbocyanine iodide, 98%
3,3'-Diethyloxacarbocyanine iodide, 98%
3,3'-Diethyloxacarbocyanine iodide, 98%
CAS: 905-96-4 | C21H22IN2O2+ | 461.323 g/mol
Have Questions?
Catalog NumberQuantity
ALFH55607.031 g
Catalog number ALFH55607.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material3,3'-Diethyloxacarbocyanine iodide
CAS905-96-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c