2-Bromomethyl-6-methylpyridine, 97%
2-Bromomethyl-6-methylpyridine, 97%
2-Bromomethyl-6-methylpyridine, 97%
2-Bromomethyl-6-methylpyridine, 97%
Thermo Scientific Chemicals
2-Bromomethyl-6-methylpyridine, 97%
CAS: 68470-59-7 | C7H8BrN | 186.05 g/mol
Have Questions?
Catalog NumberQuantity
ALFH55438.031 g
Catalog number ALFH55438.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromomethyl-6-methylpyridine
CAS68470-59-7
Health Hazard 1H302-H315-H318-H335
Health Hazard 2GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c