2-Amino-5-bromobenzonitrile, 96%
2-Amino-5-bromobenzonitrile, 96%
2-Amino-5-bromobenzonitrile, 96%
2-Amino-5-bromobenzonitrile, 96%
Thermo Scientific Chemicals
2-Amino-5-bromobenzonitrile, 96%
CAS: 39263-32-6 | C7H5BrN2 | 197.04 g/mol
Have Questions?
Catalog NumberQuantity
ALFH55217.031 g
Catalog number ALFH55217.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-5-bromobenzonitrile
CAS39263-32-6
Health Hazard 1H302+H312+H332-H315-H319-H335-H500
Health Hazard 2GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
2-Amino-5-bromobenzonitrile can react with Oxalic acid dimethyl ester to get 5-Bromo-N-methylanthranilonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-bromobenzonitrile can react with Oxalic acid dimethyl ester to get 5-Bromo-N-methylanthranilonitrile.
Solubility
Soluble in methanol.
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Edward C. Taylor.; Robert J. Knopf.; Alan L. Borror. The Dimerization of 2-Amino-5-nitrobenzonitrile. J. Am. Chem. Soc. 1960, 82, (12), 3152-3157.
  2. Zhaoyan Zheng.; Howard Alper. Palladium-Catalyzed Cyclocarbonylation of o-Iodoanilines with Imidoyl Chlorides to Produce Quinazolin-4(3H)-ones. Org. Lett. 2008, 10, (5), 829-832.