4-Bromo-N-methylbenzylamine, 97%
4-Bromo-N-methylbenzylamine, 97%
4-Bromo-N-methylbenzylamine, 97%
4-Bromo-N-methylbenzylamine, 97%
Thermo Scientific Chemicals
4-Bromo-N-methylbenzylamine, 97%
CAS: 699-03-6 | C8H10BrN | 200.079 g/mol
Have Questions?
Catalog NumberQuantity
ALFH54506.065 g
Catalog number ALFH54506.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Bromo-N-methylbenzylamine
CAS699-03-6
Health Hazard 1H302-H319
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c