4-Isobutylphenylmagnesium bromide, 0.5M in 2-MeTHF
4-Isobutylphenylmagnesium bromide, 0.5M in 2-MeTHF
4-Isobutylphenylmagnesium bromide, 0.5M in 2-MeTHF
4-Isobutylphenylmagnesium bromide, 0.5M in 2-MeTHF
Thermo Scientific Chemicals
4-Isobutylphenylmagnesium bromide, 0.5M in 2-MeTHF
CAS: 863780-45-4 | C10H13BrMg | 237.423 g/mol
Have Questions?
Catalog NumberQuantity
ALFH54201.AE100 mL
Catalog number ALFH54201.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or Material4-Isobutylphenylmagnesium bromide
CAS863780-45-4
Health Hazard 1H225-H302-H314-H335
Health Hazard 2GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c