cis-3-Hexene, 97%
cis-3-Hexene, 97%
cis-3-Hexene, 97%
Thermo Scientific Chemicals
cis-3-Hexene, 97%
CAS: 7642-09-3 | C6H12 | 84.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFH53485.065 g
Catalog number ALFH53485.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Materialcis-3-Hexene
CAS7642-09-3
Health Hazard 1H225-H304
Health Hazard 2GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
View more
cis-3-Hexene is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-3-Hexene is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. L.Sjoqvist; M.Shiotani; A.Lunda. The structure of trans- and cis-3-hexene radical cations stabilised in halocarbon matrices at low temperature: An ESR and MNDO/AM1 study. Chemical Physics. 1990, 141, (2-3),417-430
  2. D. van Hemelrijk; L. van den Enden; H.J.Geise. The molecular structures of cis-3-hexene and trans-3-hexene in the gas phase by electron diffraction and molecular mechanical calculations. Journal of Molecular Structure. 1981, 74, (1),123-135