Hexamethylacetone, 98%
Hexamethylacetone, 98%
Hexamethylacetone, 98%
Hexamethylacetone, 98%
Thermo Scientific Chemicals
Hexamethylacetone, 98%
CAS: 815-24-7 | C9H18O | 142.242 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH53464.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHexamethylacetone
CAS815-24-7
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more
Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Bellow, J. A.; Fang, D.; Kovacevic, N.; Martin, P. D.; Shearer, J.; Cisneros, G. A.; Groysman, S. Novel Alkoxide Cluster Topologies Featuring Rare Seesaw Geometry at Transition Metal Centers. Chem. Eur. J. 2013, 19 (37), 12225-12228.
  2. Cataldo, F.; Rosati, A.; Lilla, E.; Ursini, O. On the action of ozone at high concentration on various grades of polyethylene and certain straight chain paraffins. Polym. Degrad. Stab. 2011, 96 (5), 955-964.