2-Bromo-3-(bromomethyl)pyridine, 96%, Thermo Scientific Chemicals
2-Bromo-3-(bromomethyl)pyridine, 96%, Thermo Scientific Chemicals
2-Bromo-3-(bromomethyl)pyridine, 96%, Thermo Scientific Chemicals
2-Bromo-3-(bromomethyl)pyridine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-3-(bromomethyl)pyridine, 96%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFH52311.MD250 mg
Catalog number ALFH52311.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromo-3-(bromomethyl)pyridine
CAS94446-97-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c