3-Fluoro-4-iodopyridine, 98%, Thermo Scientific™
3-Fluoro-4-iodopyridine, 98%, Thermo Scientific™
3-Fluoro-4-iodopyridine, 98%, Thermo Scientific™
3-Fluoro-4-iodopyridine, 98%, Thermo Scientific™
3-Fluoro-4-iodopyridine, 98%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFH50068.031 g
Catalog number ALFH50068.03
Price (MYR)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Fluoro-4-iodopyridine
CAS22282-75-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.