Sand, washed
Sand, washed
Sand, washed
Thermo Scientific Chemicals
Sand, washed
CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
1000 g
Catalog number ALFH37933.0I
Price (MYR)
1,123.00
EA
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (MYR)
1,123.00
EA
Add to cart
Specifications
Boiling Point2230°C
Chemical Name or MaterialSand
Melting Point1710°C
Name NoteWashed
CAS14808-60-7
View more
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Joseph Wang; Robin Krause; Kirstin Block; Mustafa Musameh; Ashok Mulchandani; Michael J.Schoning. Flow injection amperometric detection of OP nerve agents based on an organophosphorus-hydrolase biosensor detector. Biosensors and Bioelectronics. 2003, 18, (2-3),255-260
  2. Angus I.Kingon; Jon-Paul Maria; S.K.Streiffer. Alternative dielectrics to silicon dioxide for memory and logic devices. Nature. 2000, 406, 1032-1038