1,3-Dibromoacetone, 95%
1,3-Dibromoacetone, 95%
1,3-Dibromoacetone, 95%
1,3-Dibromoacetone, 95%
Thermo Scientific Chemicals
1,3-Dibromoacetone, 95%
CAS: 816-39-7 | C3H4Br2O | 215.872 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH37325.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1,3-Dibromoacetone
CAS816-39-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.
Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
General References:
  1. Thierry Gefflaut; Marielle Lemaire; Marie-Lise Valentin; Jean Bolte. A novel efficient synthesis of dihydroxyacetone phosphate and bromoacetol phosphate for use in enzymatic aldol syntheses. J. Org. Chem. 1997, 62, (17),5920-5922
  2. Claudio F.Tormena; Matheus P.Freitas; Roberto Rittner; Raymond J.Abraham. Theoretical and infrared investigation of the conformations of 1,3-dihaloacetones. J. Phys. Chem. A 2004, 108, (24),5161-5168