Check Order Status
Heptane (HPLC), Fisher Chemical™
Element Search
Structure Search
Thermo Scientific Chemicals
Heptane (HPLC), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
ALFH350-4
4 L
Glass Bottle
Catalog number
ALFH350-4
Price (MYR)
422.00
EA
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
422.00
EA
Add to cart
Specifications
Chemical Name or Material
Heptane
Cleanup Symbol
Solvent
Melting Point
-91°C
Absorbance
0.014 AU max. at 254nm, 0.2 AU max. at 215nm, 0.75 AU max. at 200nm
CAS
142-82-5
View more