n-Heptane, 95%
n-Heptane, 95%
n-Heptane, 95%
Thermo Scientific Chemicals
n-Heptane, 95%
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFH33987.0F2500 mL
Catalog number ALFH33987.0F
Price (MYR)
616.00
EA
Add to cart
Quantity:
2500 mL
Request bulk or custom format
Price (MYR)
616.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Heptane
CAS142-82-5
Health Hazard 1H225-H304-H315-H319-H336-H373
Health Hazard 2GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
View more
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.
Solubility
Miscible with alcohol, chloroform, and ether. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area away from incompatible substances.
RUO – Research Use Only
General References:
  1. Caponetti, E.; Pedone, L.; Martino, D. C.; Panto, V.; Liveri, V. T. Synthesis, size control, and passivation of CdS nanoparticles in water/AOT-heptane microemulsions. Mater. Sci. Eng. C 2003, 23 (4), 531-539.
  2. Chica, A.; Corma, A. Hydroisomerization of Pentane, Hexane, and Heptane for Improving the Octane Number of Gasoline. J. Catal. 1999, 187 (1), 167-176.