2(1H)-Quinolinone-3-carboxaldehyde, 97%
2(1H)-Quinolinone-3-carboxaldehyde, 97%
2(1H)-Quinolinone-3-carboxaldehyde, 97%
2(1H)-Quinolinone-3-carboxaldehyde, 97%
2(1H)-Quinolinone-3-carboxaldehyde, 97%
CAS: 91301-03-0 | C10H7NO2 | 173.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFH33508.MD250 mg
Catalog number ALFH33508.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or Material2(1H)-Quinolinone-3-carboxaldehyde
CAS91301-03-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c