Oleyl alcohol, 99+%
Oleyl alcohol, 99+%
Oleyl alcohol, 99+%
Thermo Scientific Chemicals
Oleyl alcohol, 99+%
CAS: 143-28-2 | C18H36O | 268.49 g/mol
Have Questions?
Catalog NumberQuantity
ALFH33475.065 g
Catalog number ALFH33475.06
Price (MYR)
3,908.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
3,908.00
EA
Add to cart
Specifications
Chemical Name or MaterialOleyl alcohol
CAS143-28-2
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315
Causes skin irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
Oleyl alcohol is used in softening and lubrication of textile fabrics, in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams, lotions as a thickner, and in hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Oleyl alcohol is used in softening and lubrication of textile fabrics, in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams, lotions as a thickner, and in hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.
Solubility
Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
Notes
Incompatible with oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Liao, Y. P.; Ye, H.; Wang, Y. Preparation and Separation Properties of Oleyl Alcohol-Modified PDMS Membranes. Adv. Mater. Res. 2015, 1096, 470-473.
  2. Beltran, A. B.; Nisola, G. M.; Vivas, E. L.; Cho, W.; Chung, W. J. Poly (octylmethylsiloxane)/oleyl alcohol supported liquid membrane for the pervaporative recovery of 1-butanol from aqueous and ABE model solutions. J. Ind. Eng. Chem. 2013, 19 (1), 182-189.