3-Acetylquinoline, 97%
3-Acetylquinoline, 97%
3-Acetylquinoline, 97%
3-Acetylquinoline, 97%
Thermo Scientific Chemicals
3-Acetylquinoline, 97%
CAS: 33021-53-3 | C11H9NO | 171.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH33206.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Acetylquinoline
CAS33021-53-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c