4-(Trifluoromethylsulfonyl)phenylacetic acid, 98%
4-(Trifluoromethylsulfonyl)phenylacetic acid, 98%
4-(Trifluoromethylsulfonyl)phenylacetic acid, 98%
4-(Trifluoromethylsulfonyl)phenylacetic acid, 98%
Thermo Scientific Chemicals
4-(Trifluoromethylsulfonyl)phenylacetic acid, 98%
CAS: 1099597-82-6 | C9H7F3O4S | 268.206 g/mol
Have Questions?
Catalog NumberQuantity
ALFH32295.031 g
Catalog number ALFH32295.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(Trifluoromethylsulfonyl)phenylacetic acid
CAS1099597-82-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
4-(Trifluoromethylsulfonyl)phenylacetic acid, is used as a primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Trifluoromethylsulfonyl)phenylacetic acid, is used as a primary and secondary intermediate.
Solubility
Slightly soluble in water, soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. A Aramini.; MR Sablone.; G Bianchini.; A Amore.;M Fanì. Facile one-pot preparation of 2-arylpropionic and arylacetic acids from cyanohydrins by treatment with aqueous HI. Tetrahedron..200965 (10), 2015-2021.
  2. E Valeur.; M Bradley. Amide bond formation: beyond the myth of coupling reagents. Chemical Society Reviews.200938 (1), 606-631.