Hydrogen chloride, 5 to 6M soln. in 2-propanol, pure
Hydrogen chloride, 5 to 6M soln. in 2-propanol, pure
Hydrogen chloride, 5 to 6M soln. in 2-propanol, pure
Hydrogen chloride, 5 to 6M soln. in 2-propanol, pure
Thermo Scientific Chemicals
Hydrogen chloride, 5 to 6M soln. in 2-propanol, pure
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFH31519.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Health Hazard 1H225-H314-H332-H335-H336
Health Hazard 2GHS H Statement
H225-H314-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Health Hazard 3P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
View more
Hydrogen chloride, 5 to 6M soln. in 2-propanol may be used in measuring their conductance at 250. It is used in the manufacturing of pharmaceuticals to get a hydrochloride salt. For example, ranitidine base is converted to ranitidine hydrochloride, which is used as anti ulcerative drug.