Methyl elaidate, 95%, Thermo Scientific™
Methyl elaidate, 95%, Thermo Scientific™
Methyl elaidate, 95%, Thermo Scientific™
Methyl elaidate, 95%, Thermo Scientific™
Methyl elaidate, 95%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFH30458.031 g
Catalog number ALFH30458.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl elaidate
CAS1937-62-8
Melting Point9°C to 10°C
Recommended StorageAmbient temperatures
Density0.871
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.