1-n-Butyl-3-methylimidazolium methyl sulfate, 99%
1-n-Butyl-3-methylimidazolium methyl sulfate, 99%
1-n-Butyl-3-methylimidazolium methyl sulfate, 99%
1-n-Butyl-3-methylimidazolium methyl sulfate, 99%
Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium methyl sulfate, 99%
CAS: 401788-98-5 | C9H18N2O4S | 250.31 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27754.1850 g
Catalog number ALFH27754.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1-n-Butyl-3-methylimidazolium methyl sulfate
CAS401788-98-5
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c