Chlorobenzene, HPLC Grade, 99+%
Chlorobenzene, HPLC Grade, 99+%
Actual product may vary
Chlorobenzene, HPLC Grade, 99+%
Chlorobenzene, HPLC Grade, 99+%
Thermo Scientific Chemicals
Chlorobenzene, HPLC Grade, 99+%
CAS: 108-90-7 | C{6}H{5}Cl
Have Questions?
Catalog NumberQuantity
ALFH27688.AU1000 mL
Catalog number ALFH27688.AU
Price (MYR)
1,029.00
EA
Add to cart
Quantity:
1000 mL
Price (MYR)
1,029.00
EA
Add to cart
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point-45°C
CAS108-90-7
Boiling Point132°C
Quantity1000 mL
Molecular FormulaC{6}H{5}Cl
Percent Purity99+%
Unit SizeEA