Tetramethylammonium acetate, tech. 90%
Tetramethylammonium acetate, tech. 90%
Tetramethylammonium acetate, tech. 90%
Tetramethylammonium acetate, tech. 90%
Thermo Scientific Chemicals
Tetramethylammonium acetate, tech. 90%
CAS: 10581-12-1 | C6H15NO2 | 133.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27659.065 g
Catalog number ALFH27659.06
Price (MYR)
499.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
499.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetramethylammonium acetate
CAS10581-12-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only
General References:
  1. Popp, S.; Bösmann, A.; Wölfel, R.; Wasserscheid, P. Screening of Ionic Liquid/H2O Working Pairs for Application in Low Temperature Driven Sorption Heat Pump Systems. ACS Sustainable Chem. Eng. 2015, 3 (4), 750-757.
  2. Okamura, T. A.; Furuya, R.; Onitsuka, K. Synthesis and structures of soluble magnesium and zinc carboxylates containing intramolecular NH···O hydrogen bonds in nonpolar solvents. Dalton Trans. 2015, 44 (16), 7512-7523.