4-Mercaptophenylacetic acid, 97%
4-Mercaptophenylacetic acid, 97%
4-Mercaptophenylacetic acid, 97%
4-Mercaptophenylacetic acid, 97%
Thermo Scientific Chemicals
4-Mercaptophenylacetic acid, 97%
CAS: 39161-84-7 | C8H8O2S | 168.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27658.065 g
Catalog number ALFH27658.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Mercaptophenylacetic acid
CAS39161-84-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
An useful synthetic intermediate for the preparation of novel anti inflammatory agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
An useful synthetic intermediate for the preparation of novel anti inflammatory agents.
Solubility
Slightly soluble in water. Soluble in Dichloromethane, Ethanol and Methanol
Notes
Air sensitive. Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only