1-Cyclopropyl-4-piperidone, 98%
1-Cyclopropyl-4-piperidone, 98%
1-Cyclopropyl-4-piperidone, 98%
1-Cyclopropyl-4-piperidone, 98%
Thermo Scientific Chemicals
1-Cyclopropyl-4-piperidone, 98%
CAS: 62813-01-8 | C8H13NO | 139.20 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27495.065 g
Catalog number ALFH27495.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Cyclopropyl-4-piperidone
CAS62813-01-8
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC8H13NO