4-Iodobiphenyl, 97%
4-Iodobiphenyl, 97%
4-Iodobiphenyl, 97%
Thermo Scientific Chemicals
4-Iodobiphenyl, 97%
CAS: 1591-31-7 | C12H9I | 280.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH27453.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Iodobiphenyl
CAS1591-31-7
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point110°C to 114°C
View more
4-Iodobiphenyl is used as a starting material for the preparation of p-terphenyl derivatives. It is also used in Suzuki reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Iodobiphenyl is used as a starting material for the preparation of p-terphenyl derivatives. It is also used in Suzuki reaction.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only
General References:
  1. Okoye, N. H.; de Silva, U. K.; Wengatz, J. A.; Lapitsky, Y. Photodirected assembly of polyelectrolyte complexes. Polymer 2015, 60, 69-76.
  2. Liu, W.; Xu, L. Ligand-controlled chemoselectivity in potassium tert-amylate-promoted direct C-H arylation of unactivated benzene with aryl halides. Tetrahedron 2015, 71 (30), 4974-4981.