N-Fmoc-S-trityl-L-cysteine, 95%
N-Fmoc-S-trityl-L-cysteine, 95%
N-Fmoc-S-trityl-L-cysteine, 95%
N-Fmoc-S-trityl-L-cysteine, 95%
Thermo Scientific Chemicals
N-Fmoc-S-trityl-L-cysteine, 95%
CAS: 103213-32-7 | C37H30NO4S | 584.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFH27363.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Fmoc-S-trityl-L-cysteine
CAS103213-32-7
Melting Point171°C to 173°C
Recommended StorageKeep cold
TSCANo
View more
It is potentially useful for proteomics studies, and solid phase peptide synthesis techniques. Cysteine is versatile amino acid involved with many biological processes, including the formation of disulfide bonds - a critical component of protein structure. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is potentially useful for proteomics studies, and solid phase peptide synthesis techniques. Cysteine is versatile amino acid involved with many biological processes, including the formation of disulfide bonds - a critical component of protein structure. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function.
Solubility
Insoluble in water. Soluble in most organic solvents.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Laiping Xie, et al. Cellular Effects of Small Molecule PTP1B Inhibitors on Insulin Signaling.Biochemistry.,2003,42(44), 12792-12804.
  2. Shu-Li You, et al. Total synthesis of didmolamides A and B.Tetrahedron Lett.,2005,46(15), 2567-2570.