1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%
1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%
1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%
1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%
Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%
CAS: 412009-61-1 | C6H12N2O4S | 208.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27232.30250 g
Catalog number ALFH27232.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Ethyl-3-methylimidazolium hydrogen sulfate
CAS412009-61-1
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c