Pentaphenylcyclopentadiene, 99%
Pentaphenylcyclopentadiene, 99%
Pentaphenylcyclopentadiene, 99%
Pentaphenylcyclopentadiene, 99%
Thermo Scientific Chemicals
Pentaphenylcyclopentadiene, 99%
CAS: 2519-10-0 | C35H26 | 446.593 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27223.031 g
Catalog number ALFH27223.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPentaphenylcyclopentadiene
CAS2519-10-0
Melting Point254°C to 257°C
Recommended StorageAmbient temperatures
TSCANo
View more
1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency, in the preparation of monodisperse and highly fluorescent silica nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency, in the preparation of monodisperse and highly fluorescent silica nanoparticles.
Solubility
Soluble in pyridine 5 mg/ml.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Ohmori Y, et al. Enhancement of electroluminescence efficiency in poly (3-alkylthiophene) by doping of dye molecules.Synthetic Metals.,1997,85(1), 1241-42.
  2. Mu X, et al. A facile and general approach for the synthesis of fluorescent silica nanoparticles doped with inert dyes.Chin. Sci. Bull.,2011,756(31), 3242-46.