2,4-Dimethoxybenzylamine, 98%
2,4-Dimethoxybenzylamine, 98%
2,4-Dimethoxybenzylamine, 98%
2,4-Dimethoxybenzylamine, 98%
Thermo Scientific Chemicals
2,4-Dimethoxybenzylamine, 98%
CAS: 20781-20-8 | C9H13NO2 | 167.208 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27140.065 g
Catalog number ALFH27140.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4-Dimethoxybenzylamine
CAS20781-20-8
Health Hazard 1H314
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c