2-Chloro-6-methoxyquinoline-3-carboxaldehyde, 99%
2-Chloro-6-methoxyquinoline-3-carboxaldehyde, 99%
2-Chloro-6-methoxyquinoline-3-carboxaldehyde, 99%
2-Chloro-6-methoxyquinoline-3-carboxaldehyde, 99%
Thermo Scientific Chemicals
2-Chloro-6-methoxyquinoline-3-carboxaldehyde, 99%
CAS: 73568-29-3 | C11H8ClNO2 | 221.64 g/mol
Have Questions?
Catalog NumberQuantity
ALFH26973.065 g
Catalog number ALFH26973.06
Price (MYR)
562.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
562.00
EA
Add to cart
Specifications
Chemical Name or Material2-Chloro-6-methoxyquinoline-3-carboxaldehyde
CAS73568-29-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only
General References:
  1. Subashini,; S. Mohana Roopan,; F. nawaz khan. Synthesis and Free Radical Scavenging Property of some Quinoline Derivatives. J. Chil. Chem. Soc. 2010, 55 (3),317-319.
  2. F. N. Khan,; R. Subashini,; S. M. Roopan,; V. R. Hathwar,;S. W. Ng. 2-Chloro-6-methyl­quinoline-3-carbaldehyde . Journal name. year of publication , volume issue , pg no.