Agar, plant cell culture tested
Agar, plant cell culture tested
Thermo Scientific Chemicals
Agar, plant cell culture tested
Used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFH26724.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Recommended StorageAmbient temperatures
EINECS Number232-658-1
FormPowder
Packaging TypePlastic Bottle
Quantity500 g
View more
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.