4-n-Octylphenol, 99%
4-n-Octylphenol, 99%
4-n-Octylphenol, 99%
4-n-Octylphenol, 99%
Thermo Scientific Chemicals
4-n-Octylphenol, 99%
CAS: 1806-26-4 | C14H22O | 206.33 g/mol
Have Questions?
Catalog NumberQuantity
ALFH26396.031 g
Catalog number ALFH26396.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material4-n-Octylphenol
CAS1806-26-4
Health Hazard 1H314
Health Hazard 2GHS H Statement
H301-H314-H318-H361
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Shigeru Kohtani.; Masaya Koshiko.; Akihiko Kudo.; Kunihiro Tokumura.; Yasuhito Ishigaki.; Akira Toriba.; Kazuichi Hayakawa.; Ryoichi Nakagaki. Photodegradation of 4-alkylphenols using BiVO4 photocatalyst under irradiation with visible light from a solar simulator. Applied Catalysis B: Environmental. 2003, 46 (3), 573-586.
  2. T. Tanizaki.; K. Kadokami.; R. Shinohara. Bulletin of Environmental Contamination and Toxicology. Bulletin of Environmental Contamination and Toxicology. 2002, 68 (5), 732-739.