3,4-Difluorophenylhydrazine hydrochloride, 98%, Thermo Scientific™
3,4-Difluorophenylhydrazine hydrochloride, 98%, Thermo Scientific™
3,4-Difluorophenylhydrazine hydrochloride, 98%, Thermo Scientific™
3,4-Difluorophenylhydrazine hydrochloride, 98%, Thermo Scientific™
3,4-Difluorophenylhydrazine hydrochloride, 98%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFH26186.065 g
Catalog number ALFH26186.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3,4-Difluorophenylhydrazine hydrochloride
CAS875664-54-3
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.