meso-Tetrakis(4-methoxyphenyl)porphine, 95%
meso-Tetrakis(4-methoxyphenyl)porphine, 95%
meso-Tetrakis(4-methoxyphenyl)porphine, 95%
meso-Tetrakis(4-methoxyphenyl)porphine, 95%
Thermo Scientific Chemicals
meso-Tetrakis(4-methoxyphenyl)porphine, 95%
CAS: 22112-78-3 | C48H38N4O4 | 734.856 g/mol
Have Questions?
Catalog NumberQuantity
ALFH25946.031 g
Catalog number ALFH25946.03
Price (MYR)
558.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
558.00
EA
Add to cart
Specifications
Chemical Name or Materialmeso-Tetrakis(4-methoxyphenyl)porphine
CAS22112-78-3
Recommended StorageKeep cold
EINECS Number000-000-0
TSCANo