2-Amino-4-chlorothiophenol, 96%
2-Amino-4-chlorothiophenol, 96%
2-Amino-4-chlorothiophenol, 96%
2-Amino-4-chlorothiophenol, 96%
Thermo Scientific Chemicals
2-Amino-4-chlorothiophenol, 96%
CAS: 1004-00-8 | C6H6ClNS | 159.631 g/mol
Have Questions?
Catalog NumberQuantity
ALFB25640.065 g
Catalog number ALFB25640.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-4-chlorothiophenol
CAS1004-00-8
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c