Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate
Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate
Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate
Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate
Thermo Scientific Chemicals
Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate
CAS: 7681-38-1 | HNaO4S | 120.05 g/mol
Have Questions?
Catalog NumberQuantity
ALFB25587.0B1000 g
Catalog number ALFB25587.0B
Price (MYR)
488.00
EA
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (MYR)
488.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium hydrogen sulfate
Melting Point177°C to 180°C
Name Noteanhydrous
CAS7681-38-1
Health Hazard 1H318