1-Methylbenzimidazole, 97%
1-Methylbenzimidazole, 97%
1-Methylbenzimidazole, 97%
1-Methylbenzimidazole, 97%
Thermo Scientific Chemicals
1-Methylbenzimidazole, 97%
CAS: 1632-83-3 | C8H8N2 | 132.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFB25472.1425 g
Catalog number ALFB25472.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Methylbenzimidazole
CAS1632-83-3
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
1-Methylbenzimidazole is the suitable reagent used as electrolyte additive for the dye-sensitized solar cells and solid-state dye-sensitized solar cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methylbenzimidazole is the suitable reagent used as electrolyte additive for the dye-sensitized solar cells and solid-state dye-sensitized solar cells.
Solubility
Soluble in methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Gerrit Boschloo; Henrik Lindström; Eva Magnusson; Anna Holmberg; Anders Hagfeldt. Optimization of dye-sensitized solar cells prepared by compression method. Journal of Photochemistry and Photobiology A: Chemistry. 2002, 148 (1-3), 11-15.
  2. Nikolaos Balis; Vassilios Dracopoulos; Maria Antoniadou; Panagiotis Lianos. Solid-state dye-sensitized solar cells made of multilayer nanocrystalline titania and poly(3-hexylthiophene). Journal of Photochemistry and Photobiology A: Chemistry. 2010, 214 (1), 69-73.