5-Amino-2-chloropyridine, 98%
5-Amino-2-chloropyridine, 98%
5-Amino-2-chloropyridine, 98%
5-Amino-2-chloropyridine, 98%
5-Amino-2-chloropyridine, 98%
CAS: 5350-93-6 | C5H5ClN2 | 128.56 g/mol
Have Questions?
Catalog NumberQuantity
ALFB25336.031 g
Catalog number ALFB25336.03
Price (MYR)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Amino-2-chloropyridine
CAS5350-93-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c