1,3-Di-o-tolylguanidine, 99%
1,3-Di-o-tolylguanidine, 99%
1,3-Di-o-tolylguanidine, 99%
1,3-Di-o-tolylguanidine, 99%
Thermo Scientific Chemicals
1,3-Di-o-tolylguanidine, 99%
CAS: 97-39-2 | C15H17N3 | 239.322 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALFB25127.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material1,3-Di-o-tolylguanidine
CAS97-39-2
Health Hazard 1H301
Health Hazard 2GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P301+P310-P330-P501c