Nalidixic acid, 99%
Nalidixic acid, 99%
Nalidixic acid, 99%
Thermo Scientific Chemicals
Nalidixic acid, 99%
CAS: 389-08-2 | C12H12N2O3 | 232.239 g/mol
Have Questions?
Catalog NumberQuantity
ALFB25096.065 g
Catalog number ALFB25096.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNalidixic acid
CAS389-08-2
Health Hazard 1H317-H334-H336-H341
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
View more
Nalidixic acid is a synthetic 1,8-naphthyridine antimicrobial agent. It is also used as an inhibitor of bacterial DNA gyrase. It plays an important role in the treatment of urinary tract infections caused by susceptible gram-negative microorganisms, including the majority of E. Coli, Enterobacter species, Klebsiella species and Proteus species.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nalidixic acid is a synthetic 1,8-naphthyridine antimicrobial agent. It is also used as an inhibitor of bacterial DNA gyrase. It plays an important role in the treatment of urinary tract infections caused by susceptible gram-negative microorganisms, including the majority of E. Coli, Enterobacter species, Klebsiella species and Proteus species.
Solubility
Soluble in diethyl ether, acetone, methylene chloride, chloroform and methanol. Insoluble in water.
Notes
Light sensitive and air sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Chintagari, S.; Hung, Y. C.; Hamanaka, D. Resistance of various STEC strains and serogroups to UV radiation and effect of nalidixic acid adaptation. Food Control 2015, 50, 313-319.
  2. Dwivedi, G. R.; Maurya, A.; Yadav, D. K.; Khan, F.; Darokar, M. P.; Srivastava, S. K. Drug Resistance Reversal Potential of Ursolic Acid Derivatives against Nalidixic Acid- and Multidrug-resistant Escherichia coli. Chem. Biol. Drug Des. 2015, 86 (3), 272-283.