Chlorocyclopentane, 99%
Chlorocyclopentane, 99%
Chlorocyclopentane, 99%
Chlorocyclopentane, 99%
Thermo Scientific Chemicals
Chlorocyclopentane, 99%
CAS: 930-28-9 | C5H9Cl | 104.577 g/mol
Have Questions?
Catalog NumberQuantity
ALFB25023.22100 g
Catalog number ALFB25023.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialChlorocyclopentane
CAS930-28-9
Health Hazard 1H225-H315-H319
Health Hazard 2GHS H Statement
H225
Highly flammable liquid and vapour.
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
View more
Chlorocyclopentane was used in the preparation of diastereomeric activation products. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorocyclopentane was used in the preparation of diastereomeric activation products. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water. 760 mg/L.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only
General References:
  1. Andrew J Vetter; Ryan D Rieth; William W Brennessel; William D Jones. Selective C-H activation of haloalkanes using a rhodiumtrispyrazolylborate complex. Journal of the American Chemical Society.2009, 131, (30), 10742-10752.
  2. Glen A. Russell; Akihiko. Ito. Photochlorination of Bromocyclopentane and Chlorocyclopentane. Journal of the American Chemical Society.1963, 85, (19), 2983-2988.