3,4,5-Trihydroxybenzoic acid, 98%
3,4,5-Trihydroxybenzoic acid, 98%
3,4,5-Trihydroxybenzoic acid, 98%
3,4,5-Trihydroxybenzoic acid, 98%
Thermo Scientific Chemicals
3,4,5-Trihydroxybenzoic acid, 98%
CAS: 149-91-7 | C7H6O5 | 170.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFB24887.30
Price (MYR)
805.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
805.00
EA
Add to cart
Specifications
Chemical Name or Material3,4,5-Trihydroxybenzoic acid
Name Noteanhydrous
CAS149-91-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.
Solubility
Soluble in water, alcohol, ether, glycerol and acetone. Insoluble in benzene, chloroform and petroleum ether.
Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents, strong bases, acid anhydrides, acid chlorides, metals.
RUO – Research Use Only
General References:
  1. Pisani, O.; Frey, S. D.; Simpson, A. J.; Simpson, M. J. Soil warming and nitrogen deposition alter soil organic matter composition at the molecular-level. Biogeochemistry 2015, 123 (3), 391-409.
  2. Diamanti, J.; Francesca, B.; Franco, C.; Clara, B.; Annalisa, B.; Francesca, G.; Maurizio, B.; Bruno, M. Physico-chemical characteristics of thermally processed purée from different strawberry genotypes. J. Food Compos. Anal. 2015, 43, 106-118.