Dimidium bromide, 95%
Dimidium bromide, 95%
Dimidium bromide, 95%
Dimidium bromide, 95%
Thermo Scientific Chemicals
Dimidium bromide, 95%
CAS: 518-67-2 | C20H18BrN3 | 380.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
0.25 g
Catalog number ALFB24818.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDimidium bromide
CAS518-67-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Dimidium bromide acts as an inhibitor of L-cell growth. It is also used as a fluorescent probe for nucleic acids. It is also useful in the determination of cationic surfactants. It is also used as an intercalating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimidium bromide acts as an inhibitor of L-cell growth. It is also used as a fluorescent probe for nucleic acids. It is also useful in the determination of cationic surfactants. It is also used as an intercalating agent.
Solubility
Soluble in water.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Makarov, N. M.; Kulapin, E. G. New potentiometric screen-printed sensors for determination of homologous sodium alkylsulfates . Sens. Actuators, B 2015, 210, 817-824.
  2. Yang, Y. J.; Kelkar, A. V.; Zhu, X.; Bai, G.; Ng, H. T.; Corti, D. S.; Franses, E. I. Effect of sodium dodecylsulfate monomers and micelles on the stability of aqueous dispersions of titanium dioxide pigment nanoparticles against agglomeration and sedimentation. J. Colloid Interface Sci. 2015, 450, 434-445.