1-Methylcyclopentanol, 98%
1-Methylcyclopentanol, 98%
1-Methylcyclopentanol, 98%
1-Methylcyclopentanol, 98%
1-Methylcyclopentanol, 98%
CAS: 1462-03-9 | C6H12O | 100.161 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24744.1425 g
Catalog number ALFB24744.14
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Methylcyclopentanol
CAS1462-03-9
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P240-P241-P280-P370+P378q